3,5-Bis(trifluormethyl)benzamid

Lieferant: FLUOROCHEM

Synonyme: 3,5-Bis(trifluormethyl)benzoesäureamid

006001-100G 006001-25G 006001-5G
FLUO006001-100GEA 515 EUR
FLUO006001-100G FLUO006001-25G FLUO006001-5G
3,5-Bis(trifluormethyl)benzamid
3,5-Bis(trifluormethyl)benzamid
Formel: C₉H₅F₆NO
Molecular Weight: 257,14 g/mol
MDL: MFCD00015511
CAS-Nummer: 22227-26-5
EINECS: 244-849-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR