3,5-Bis(trifluormethyl)benzamidin Hydrochlorid
Lieferant: Thermo Scientific
Synonyme:
3,5-Bis(trifluormethyl)benzoesäureamidin Hydrochlorid, 3,5-Bis(trifluormethyl)phenylamidin Hydrochlorid
RF00749DA
RF00749EA
MAYBRF00749DAEA
0
EUR
MAYBRF00749DA
MAYBRF00749EA
3,5-Bis(trifluormethyl)benzamidin Hydrochlorid
3,5-Bis(trifluormethyl)benzamidin Hydrochlorid
Formel:
C₉H₇ClF₆N₂ |
MDL:
MFCD00276604 CAS-Nummer: 97603-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...