3,5-Bis(trifluormethyl)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3,5-Bis(trifluormethyl)benzolsulfochlorid, 3,5-Bis(trifluormethyl)benzol-1-sulfonylchlorid, 3,5-Bis(trifluormethyl)benzolsulfonsäurechlorid
006003-5G
006003-25G
006003-100G
FLUO006003-5GEA
55.4
EUR
FLUO006003-5G
FLUO006003-25G
FLUO006003-100G
3,5-Bis(trifluormethyl)benzolsulfonylchlorid
3,5-Bis(trifluormethyl)benzolsulfonylchlorid
Formel:
C₈H₃ClF₆O₂S Schmelzpunkt: 36…38 °C |
MDL:
MFCD00014725 CAS-Nummer: 39234-86-1 EINECS: 254-371-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...