3,5-Bis(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3,5-Bis(trifluormethyl)benzolsulfochlorid, 3,5-Bis(trifluormethyl)benzol-1-sulfonylchlorid, 3,5-Bis(trifluormethyl)benzolsulfonsäurechlorid
M-Clarity™ Qualitätsstufe = MQ200
519367-1G
519367-5G
SIAL519367-1GEA
25
EUR
SIAL519367-1G
SIAL519367-5G
3,5-Bis(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®
3,5-Bis(trifluormethyl)benzolsulfonylchlorid
Formel:
C₈H₃ClF₆O₂S Molecular Weight: 312,62 g/mol Schmelzpunkt: 36…38 °C |
MDL:
MFCD00014725 CAS-Nummer: 39234-86-1 EINECS: 254-371-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...