3,5-Bis(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3,5-Bis(trifluormethyl)phenyl)metaborsäure, 3,5-Bis(trifluormethyl)benzolboronsäure, Dihydroxy(3,5-bis(trifluormethyl)phenyl)boran

BB-2640-100G BB-2640-5G BB-2640-25G
COBBBB-2640-100GEA 308 EUR
COBBBB-2640-100G COBBBB-2640-5G COBBBB-2640-25G
3,5-Bis(trifluormethyl)phenylboronsäure
3,5-Bis(trifluormethyl)phenylboronsäure
Formel: C₈H₅BF₆O₂
Molecular Weight: 257.93 g/mol
MDL: MFCD00051850
CAS-Nummer: 73852-19-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR