Methyl-3,5-diaminobenzoat ≥99%
Lieferant: Thermo Scientific
Synonyme:
3,5-Diaminobenzoesäuremethylester
L10848.09
L10848.18
L10848.09EA
62.8
EUR
L10848.09
L10848.18
Methyl-3,5-diaminobenzoat ≥99%
Methyl-3,5-diaminobenzoat
Purity: 99%
CAS No.: 1949-55-9
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Odour: Odourless
Melting Point: 131-134°
Beilstein Reference: 2363309
MDL No.: MFCD00041433
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,18 g/mol Siedepunkt: 319 °C (1013 hPa) Schmelzpunkt: 131…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041433 CAS-Nummer: 1949-55-9 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...