3,5-Dibrombenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

OS-7526-25G OS-7526-100G OS-7526-5G
COBBOS-7526-25GEA 58.4 EUR
COBBOS-7526-25G COBBOS-7526-100G COBBOS-7526-5G
3,5-Dibrombenzoesäure
3,5-Dibromobenzoic acid
Formel: C₇H₄Br₂O₂
Molecular Weight: 279,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051758
CAS-Nummer: 618-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR