3,5-Dibrompyridin

Lieferant: COMBI-BLOCKS
Achtung

OT-0355-25G OT-0355-1KG OT-0355-100G
COBBOT-0355-25GEA 37.5 EUR
COBBOT-0355-25G COBBOT-0355-1KG COBBOT-0355-100G
3,5-Dibrompyridin
3,5-Dibromopyridine
Formel: C₅H₃Br₂N
Molecular Weight: 236,89 g/mol
Siedepunkt: 222 °C (1013 hPa)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014634
CAS-Nummer: 625-92-3
EINECS: 210-916-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR