3,5-Dichlorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

BB-2269-5G BB-2269-25G BB-2269-100G
COBBBB-2269-5GEA 38.2 EUR
COBBBB-2269-5G COBBBB-2269-25G COBBBB-2269-100G
3,5-Dichlorphenylboronsäure
3,5-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190,82 g/mol
MDL: MFCD00051935
CAS-Nummer: 67492-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR