3,5-Difluorbenzolsulfonamid
Lieferant: FLUOROCHEM
Synonyme:
3,5-Difluorbenzolsulfonsäureamid
013812-5G
013812-25G
013812-1G
FLUO013812-5GEA
86.3
EUR
FLUO013812-5G
FLUO013812-25G
FLUO013812-1G
3,5-Difluorbenzolsulfonamid
3,5-Difluorbenzolsulfonamid
Formel:
C₆H₅F₂NO₂S Molecular Weight: 193,17 g/mol |
MDL:
MFCD02091379 CAS-Nummer: 140480-89-3 UN: 3144 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...