3,5-Difluorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: (3,5-Difluorophenyl)metaboric acid, Dihydroxy(3,5-difluorophenyl)borane, 3,5-Difluorobenzeneboronic acid

BB-2608-25G BB-2608-5G BB-2608-100G
COBBBB-2608-25GEA 104 EUR
COBBBB-2608-25G COBBBB-2608-5G COBBBB-2608-100G
3,5-Difluorphenylboronsäure
3,5-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
Molecular Weight: 157,91 g/mol
MDL: MFCD01318138
CAS-Nummer: 156545-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR