3,5-Difluorpyridin

Lieferant: COMBI-BLOCKS

OS-7631-5G OS-7631-25G OS-7631-1G
COBBOS-7631-5GEA 112 EUR
COBBOS-7631-5G COBBOS-7631-25G COBBOS-7631-1G
3,5-Difluorpyridin
3,5-Difluoropyridine
Formel: C₅H₃F₂N
Molecular Weight: 115.08 g/mol
Siedepunkt: 92…93 °C (1013 hPa)
Dichte: 1.268
MDL: MFCD03412222
CAS-Nummer: 71902-33-5
UN: 1993
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR