3,5-Dimethylphenylboronsäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid
BB-2787-5G
BB-2787-25G
BB-2787-100G
COBBBB-2787-5GEA
28.5
EUR
COBBBB-2787-5G
COBBBB-2787-25G
COBBBB-2787-100G
3,5-Dimethylphenylboronsäure
3,5-Dimethylphenylboronsäure
Formel:
C₈H₁₁BO₂ Molecular Weight: 149,98 g/mol |
MDL:
MFCD00185689 CAS-Nummer: 172975-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...