3-Amino-2-brom-6-chlorpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
2-Brom-6-chlor-3-pyridylamin
HC-6875-5G
HC-6875-25G
HC-6875-1G
COBBHC-6875-5GEA
323
EUR
COBBHC-6875-5G
COBBHC-6875-25G
COBBHC-6875-1G
3-Amino-2-brom-6-chlorpyridin
3-Amino-2-bromo-6-chloropyridine
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,46 g/mol |
MDL:
MFCD11504927 CAS-Nummer: 1050501-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...