3-Amino-2-naphthol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Amino-2-hydroxynaphthalin

164267-1G 164267-5G
SIAL164267-1GEA 74.9 EUR
SIAL164267-1G SIAL164267-5G
3-Amino-2-naphthol, Sigma-Aldrich®
3-Amino-2-naphthol
Formel: C₁₀H₉NO
Molecular Weight: 159,19 g/mol
MDL: MFCD00004113
CAS-Nummer: 5417-63-0
EINECS: 226-519-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR