3-Amino-2-naphthol

Lieferant: CHEMPUR

Synonyme: 3-Amino-2-hydroxynaphthalin

RJC00627.250 RJC00627.1
CHMPRJC00627.250EA 0 EUR
CHMPRJC00627.250 MAYBRJC00627.1
3-Amino-2-naphthol
3-Amino-2-naphthol
Formel: C₁₀H₉NO
Molecular Weight: 159.19 g/mol
MDL: MFCD00004113
CAS-Nummer: 5417-63-0
EINECS: 226-519-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR