3-Amino-2-naphthoesäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Amino-2-naphthalincarbonsäure, 3-Aminonaphthalin-2-carbonsäure
OS-7477-25G
OS-7477-1G
OS-7477-100G
OS-7477-5G
COBBOS-7477-25GEA
0
EUR
COBBOS-7477-25G
COBBOS-7477-1G
COBBOS-7477-100G
COBBOS-7477-5G
3-Amino-2-naphthoesäure
3-Amino-2-naphthoesäure
Formel:
C₁₁H₉NO₂ Siedepunkt: 383 °C (1013 hPa) Schmelzpunkt: 216…217 °C |
MDL:
MFCD00004115 CAS-Nummer: 5959-52-4 EINECS: 227-726-2 Merck Index: 13,00451 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...