3-Amino-5-brom-2-chlorpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-chlor-3-pyridylamin

PY-7139-5G PY-7139-25G
COBBPY-7139-5GEA 25.9 EUR
COBBPY-7139-5G COBBPY-7139-25G
3-Amino-5-brom-2-chlorpyridin
3-Amino-5-bromo-2-chloropyridine
Formel: C₅H₄BrClN₂
Molecular Weight: 207.46 g/mol
MDL: MFCD02682092
CAS-Nummer: 588729-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR