5-Amino-3-brom-2-chlorpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-2-chlor-5-pyridylamin

PY-7783-5G PY-7783-25G PY-7783-100G
COBBPY-7783-5GEA 31 EUR
COBBPY-7783-5G COBBPY-7783-25G COBBPY-7783-100G
5-Amino-3-brom-2-chlorpyridin
5-Amino-3-brom-2-chlorpyridin
Formel: C₅H₄BrClN₂
Molecular Weight: 207.46 g/mol
MDL: MFCD03840433
CAS-Nummer: 130284-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR