3-Amino-5-brombenzoesäure

Lieferant: COMBI-BLOCKS

CA-4114-25G CA-4114-5G CA-4114-1G
COBBCA-4114-25GEA 180 EUR
COBBCA-4114-25G COBBCA-4114-5G COBBCA-4114-1G
3-Amino-5-brombenzoesäure
3-Amino-5-bromobenzoic acid
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
MDL: MFCD00227745
CAS-Nummer: 42237-85-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR