3-Amino-5-brompyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
5-Brom-3-pyridylamin, 5-Brompyridin-3-amin
PY-7125-100G
PY-7125-5G
PY-7125-1G
PY-7125-25G
COBBPY-7125-100GEA
385
EUR
COBBPY-7125-100G
COBBPY-7125-5G
COBBPY-7125-1G
COBBPY-7125-25G
3-Amino-5-brompyridin
3-Amino-5-brompyridin
Formel:
C₅H₅BrN₂ Molecular Weight: 173.01 g/mol |
MDL:
MFCD01646060 CAS-Nummer: 13535-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...