3-Amino-5-nitrobenzoesäure 98%

Lieferant: Thermo Scientific
Achtung

A12508.06 A12508.14
A12508.06EA 104 EUR
A12508.06 A12508.14
3-Amino-5-nitrobenzoesäure 98%
3-Amino-5-nitrobenzoic acid

Purity: 98% CAS No.: 618-84-8 Molecular Formula: C7H6N2O4 Formula Weight: 182.14 Melting Point: 214-218° Beilstein Reference: 1457935

Formel: C₇H₆N₂O₄
Molecular Weight: 182,14 g/mol
Schmelzpunkt: 214…218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017025
CAS-Nummer: 618-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR