3-Amino-5-nitrobenzoesäure
Lieferant: FLUOROCHEM
021284-2G
021284-10G
FLUO021284-2GEA
0
EUR
FLUO021284-2G
FLUO021284-10G
3-Amino-5-nitrobenzoesäure
3-Amino-5-nitrobenzoic acid
Formel:
C₇H₆N₂O₄ Molecular Weight: 182,14 g/mol |
MDL:
MFCD00017025 CAS-Nummer: 618-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...