5-Amino-2-brompyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2-Brompyridin-5-amin, 2-Brom-5-pyridylamin

CA-4315-5G CA-4315-25G CA-4315-1G
COBBCA-4315-5GEA 38.7 EUR
COBBCA-4315-5G COBBCA-4315-25G COBBCA-4315-1G
5-Amino-2-brompyridin
5-Amino-2-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
MDL: MFCD01646116
CAS-Nummer: 13534-97-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR