3-Amino-6-chlorpyridazin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 6-Chlorpyridazin-3-amin

HC-6004-25G HC-6004-100G HC-6004-5G
COBBHC-6004-25GEA 59.1 EUR
COBBHC-6004-25G COBBHC-6004-100G COBBHC-6004-5G
3-Amino-6-chlorpyridazin
3-Amino-6-chlorpyridazin
Formel: C₄H₄ClN₃
Molecular Weight: 129,55 g/mol
MDL: MFCD00051506
CAS-Nummer: 5469-69-2
EINECS: 226-796-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR