5-Amino-2-chlorpyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2-Chlor-5-pyridylamin, 2-Chlorpyridin-5-amin
CA-4301-100G
CA-4301-25G
CA-4301-5G
COBBCA-4301-100GEA
207
EUR
COBBCA-4301-100G
COBBCA-4301-25G
COBBCA-4301-5G
5-Amino-2-chlorpyridin
5-Amino-2-chlorpyridin
Formel:
C₅H₅ClN₂ Molecular Weight: 128,56 g/mol Siedepunkt: 225 °C (1013 hPa) Schmelzpunkt: 83 °C |
MDL:
MFCD00006243 CAS-Nummer: 5350-93-6 EINECS: 226-322-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...