3-Amino-9-ethylcarbazol (AEC) 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 9-Ethylcarbazol-3-ylamin, AEC

B22529.14 B22529.22
B22529.14EA 74.9 EUR
B22529.14 B22529.22
3-Amino-9-ethylcarbazol (AEC) 95%
3-Amino-9-ethyl carbazole (AEC)

Purity: 95% CAS No.: 132-32-1 Molecular Formula: C14H14N2 Formula Weight: 210.28 Odour: Odourless Melting Point: 98-100° MDL No.: MFCD00004964

Formel: C₁₄H₁₄N₂
Molecular Weight: 210,28 g/mol
Schmelzpunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004964
CAS-Nummer: 132-32-1
EINECS: 205-055-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR