3-Amino-9-ethylcarbazol (AEC), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
9-Ethylcarbazol-3-ylamin, AEC
M-Clarity™ Qualitätsstufe = MQ200
A5754-100G
A5754-10G
A5754-50G
SIALA5754-100GEA
235
EUR
SIALA5754-100G
SIALA5754-10G
SIALA5754-50G
3-Amino-9-ethylcarbazol (AEC), Sigma-Aldrich®
3-Amino-9-ethyl carbazole (AEC)
Formel:
C₁₄H₁₄N₂ Molecular Weight: 210,28 g/mol Schmelzpunkt: 113…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004964 CAS-Nummer: 132-32-1 EINECS: 205-057-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...