3-Aminobenzolsulfonamid
Lieferant: Thermo Scientific
Synonyme:
Anilin-3-sulfonsäureamid, 3-Aminobenzolsulfonsäureamid
SB01949
SB01949DA
SB01949.10
SB01949EE
MAYBSB01949EA
0
EUR
MAYBSB01949
MAYBSB01949DA
MAYBSB01949.10
MAYBSB01949EE
3-Aminobenzolsulfonamid
3-Aminobenzolsulfonamid
Formel:
C₆H₈N₂O₂S Molecular Weight: 172,21 g/mol |
MDL:
MFCD00035781 CAS-Nummer: 98-18-0 EINECS: 202-646-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...