3-Aminophenylboronsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dihydroxy(3-aminophenyl)boran, 3-Aminobenzolboronsäure, (3-Aminophenyl)metaborsäure
H52909.03
H52909.06
H52909.14
H52909.03EA
39
EUR
H52909.03
H52909.06
H52909.14
3-Aminophenylboronsäure 98%
3-Aminophenylboronsäure
Purity: 98%
CAS No.: 30418-59-8
Molecular Formula: C6H8BNO2
Formula Weight: 136.95
Melting Point: 138-143°
MDL No.: MFCD00007755
Formel:
C₆H₈BNO₂ Molecular Weight: 136.95 g/mol Schmelzpunkt: 138…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007755 CAS-Nummer: 30418-59-8 EINECS: 250-189-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...