3-Aminophenylboronsäure Monohydrat

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(3-aminophenyl)boran Monohydrat, (3-Aminophenyl)metaborsäure Monohydrat, 3-Aminobenzolboronsäure Monohydrat

BO-2141-5G BO-2141-25G BO-2141-100G
COBBBO-2141-5GEA 0 EUR
COBBBO-2141-5G COBBBO-2141-25G COBBBO-2141-100G
3-Aminophenylboronsäure Monohydrat
3-Aminophenylboronsäure Monohydrat
Formel: C₆H₈BNO₂·1H₂O
Molecular Weight: 154.96 g/mol
MDL: MFCD00149554
CAS-Nummer: 206658-89-1
EINECS: 250-189-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR