3-Brom-2,5-dichlorpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-2,5-dichloro-pyridine

PY-7721-1G PY-7721-25G PY-7721-5G
COBBPY-7721-1GEA 0 EUR
COBBPY-7721-1G COBBPY-7721-25G COBBPY-7721-5G
3-Brom-2,5-dichlorpyridin
3-Bromo-2,5-dichloropyridine
Formel: C₅H₂BrCl₂N
Molecular Weight: 226,89 g/mol
MDL: MFCD06659484
CAS-Nummer: 138006-41-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR