3-Brom-2,5-dichlorthiophen

Lieferant: COMBI-BLOCKS

HI-1684-5G HI-1684-25G HI-1684-1G
COBBHI-1684-5GEA 76.1 EUR
COBBHI-1684-5G COBBHI-1684-25G COBBHI-1684-1G
3-Brom-2,5-dichlorthiophen
3-Bromo-2,5-dichlorothiophene
Formel: C₄HBrCl₂S
Molecular Weight: 231.93 g/mol
Siedepunkt: 89…92 °C (11 mmHg)
MDL: MFCD00041440
CAS-Nummer: 60404-18-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR