3-Brom-2,6-dichlorpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Dichloro-3-bromopyridine

PY-1227-5G PY-1227-1G
COBBPY-1227-5GEA 88.4 EUR
COBBPY-1227-5G COBBPY-1227-1G
3-Brom-2,6-dichlorpyridin
3-Bromo-2,6-dichloropyridine
Formel: C₅H₂BrCl₂N
Molecular Weight: 226,89 g/mol
MDL: MFCD06798231
CAS-Nummer: 866755-20-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR