3-Brom-2-chlor-5-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

PY-7238-1G PY-7238-5G
COBBPY-7238-1GEA 21.6 EUR
COBBPY-7238-1G COBBPY-7238-5G
3-Brom-2-chlor-5-(trifluormethyl)pyridin
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine
Formel: C₆H₂BrClF₃N
MDL: MFCD09878432
CAS-Nummer: 71701-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR