3-Brom-2-chlor-5-fluorpyridin

Lieferant: COMBI-BLOCKS

PY-7350-1G PY-7350-5G
COBBPY-7350-1GEA 38 EUR
COBBPY-7350-1G COBBPY-7350-5G
3-Brom-2-chlor-5-fluorpyridin
3-Bromo-2-chloro-5-fluoropyridine
Formel: C₅H₂BrClFN
Molecular Weight: 210.43 g/mol
MDL: MFCD03095269
CAS-Nummer: 884494-36-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR