3-Brom-2-chlor-5-nitropyridin
Lieferant: COMBI-BLOCKS
OT-0072-5G
OT-0072-25G
OT-0072-1G
OT-0072-100G
COBBOT-0072-5GEA
38
EUR
COBBOT-0072-5G
COBBOT-0072-25G
COBBOT-0072-1G
COBBOT-0072-100G
3-Brom-2-chlor-5-nitropyridin
3-Bromo-2-chloro-5-nitropyridine
Formel:
C₅H₂BrClN₂O₂ |
MDL:
MFCD00233989 CAS-Nummer: 5470-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...