3-Brom-2-chlor-6-methoxypyridin

Lieferant: COMBI-BLOCKS

PY-7361-1G PY-7361-5G
COBBPY-7361-1GEA 0 EUR
COBBPY-7361-1G COBBPY-7361-5G
3-Brom-2-chlor-6-methoxypyridin
3-Bromo-2-chloro-6-methoxypyridine
Formel: C₆H₅BrClNO
MDL: MFCD10699710
CAS-Nummer: 777931-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR