3-Brom-2-chlor-6-methylpyridin 98+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Brom-2-chlor-6-picolin, 5-Brom-6-chlor-2-picolin, 5-Brom-6-chlor-2-methylpyridin
360800010
360800050
ACRO360800010EA
33.4
EUR
ACRO360800010
ACRO360800050
3-Brom-2-chlor-6-methylpyridin 98+%
3-Bromo-2-chloro-6-methylpyridine
Formel:
C₆H₅BrClN Molecular Weight: 206.47 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095218 CAS-Nummer: 185017-72-5 |
Spezifikation Testergebnisse
Appearance | Yellow Low melting solid or liquid |
Infrared spectrum | Conforms |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...