3-Brom-2-chlor-6-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-2-chlor-6-picolin, 5-Brom-6-chlor-2-picolin, 5-Brom-6-chlor-2-methylpyridin

OS-7582-5G OS-7582-25G OS-7582-1G
COBBOS-7582-5GEA 88.6 EUR
COBBOS-7582-5G COBBOS-7582-25G COBBOS-7582-1G
3-Brom-2-chlor-6-methylpyridin
3-Bromo-2-chloro-6-methylpyridine
Formel: C₆H₅BrClN
Molecular Weight: 206,47 g/mol
MDL: MFCD03095218
CAS-Nummer: 185017-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR