3-Brom-2-chlorpyridin

Lieferant: COMBI-BLOCKS
Achtung

OS-7406-100G OS-7406-25G OS-7406-5G
COBBOS-7406-100GEA 168 EUR
COBBOS-7406-100G COBBOS-7406-25G COBBOS-7406-5G
3-Brom-2-chlorpyridin
3-Bromo-2-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
Siedepunkt: 205
Schmelzpunkt: 55
MDL: MFCD00234007
CAS-Nummer: 52200-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR