3-Brom-2-chlorthiophen

Lieferant: COMBI-BLOCKS
Achtung

OS-7540-1G OS-7540-25G OS-7540-5G
COBBOS-7540-1GEA 26.2 EUR
COBBOS-7540-1G COBBOS-7540-25G COBBOS-7540-5G
3-Brom-2-chlorthiophen
3-Bromo-2-chlorothiophene
Formel: C₄H₂BrClS
Molecular Weight: 197.48 g/mol
Siedepunkt: 195
Dichte: 1.82 g/cm³ (20 °C)
MDL: MFCD00043883
CAS-Nummer: 40032-73-3
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR