3-Brom-2-fluorbenzaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-2-fluorobenzenecarbaldehyde

CA-4741-5G CA-4741-25G CA-4741-1G
COBBCA-4741-5GEA 63.2 EUR
COBBCA-4741-5G COBBCA-4741-25G COBBCA-4741-1G
3-Brom-2-fluorbenzaldehyd
3-Bromo-2-fluorobenzaldehyde
Formel: C₇H₄BrFO
MDL: MFCD09027089
CAS-Nummer: 149947-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR