3-Brom-2-naphthol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Brom-2-hydroxynaphthalin
690120-1G
690120-5G
SIAL690120-1GEA
141
EUR
SIAL690120-1G
SIAL690120-5G
3-Brom-2-naphthol, Sigma-Aldrich®
3-Brom-2-naphthol
Formel:
C₁₀H₇BrO Molecular Weight: 223,07 g/mol |
MDL:
MFCD10000951 CAS-Nummer: 30478-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...