3-Brom-2-hydroxypyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-2-pyridinol

OS-7522-5G OS-7522-25G OS-7522-1G
COBBOS-7522-5GEA 38.5 EUR
COBBOS-7522-5G COBBOS-7522-25G COBBOS-7522-1G
3-Brom-2-hydroxypyridin
3-Brom-2-hydroxypyridin
Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03411569
CAS-Nummer: 13466-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR