3-Brom-2-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brom-2-picolin

OS-7475-5G OS-7475-25G OS-7475-1G OS-7475-100G
COBBOS-7475-5GEA 38.5 EUR
COBBOS-7475-5G COBBOS-7475-25G COBBOS-7475-1G COBBOS-7475-100G
3-Brom-2-methylpyridin
3-Brom-2-methylpyridin
Formel: C₆H₆BrN
Molecular Weight: 172,02 g/mol
MDL: MFCD00191224
CAS-Nummer: 38749-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR