3-Brom-4-chlorbenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

OS-7552-1G OS-7552-5G OS-7552-25G
COBBOS-7552-1GEA 0 EUR
COBBOS-7552-1G COBBOS-7552-5G COBBOS-7552-25G
3-Brom-4-chlorbenzoesäure
3-Bromo-4-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
MDL: MFCD00079706
CAS-Nummer: 42860-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR