3-Brom-4-methylpyridin

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 3-Bromo-4-picoline

PY-7011-25G PY-7011-1G PY-7011-5G PY-7011-100G
COBBPY-7011-25GEA 89.5 EUR
COBBPY-7011-25G COBBPY-7011-1G COBBPY-7011-5G COBBPY-7011-100G
3-Brom-4-methylpyridin
3-Brom-4-methylpyridin
Formel: C₆H₆BrN
Molecular Weight: 172.02 g/mol
Siedepunkt: 47…48 °C (0.6 mmHg)
Dichte: 1.549 g/cm³ (20 °C)
MDL: MFCD00082592
CAS-Nummer: 3430-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR