3-Brom-4-methylthiophen
Lieferant: COMBI-BLOCKS
OS-7571-5G
OS-7571-25G
OS-7571-1G
COBBOS-7571-5GEA
114
EUR
COBBOS-7571-5G
COBBOS-7571-25G
COBBOS-7571-1G
3-Brom-4-methylthiophen
3-Bromo-4-methylthiophene
Formel:
C₅H₅BrS Molecular Weight: 177.06 g/mol |
MDL:
MFCD00130084 CAS-Nummer: 30318-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...