3-Brom-5-chlor-2-fluorpyridin

Lieferant: COMBI-BLOCKS

SS-3014-1G SS-3014-25G SS-3014-5G
COBBSS-3014-1GEA 26.2 EUR
COBBSS-3014-1G COBBSS-3014-25G COBBSS-3014-5G
3-Brom-5-chlor-2-fluorpyridin
3-Bromo-5-chloro-2-fluoropyridine
Formel: C₅H₂BrClFN
Molecular Weight: 210.43 g/mol
MDL: MFCD04972409
CAS-Nummer: 884494-87-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR